8-Chloroquinoline 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
405020100 405020250
ACRO405020100EA 56.6 EUR
ACRO405020100 ACRO405020250
8-Chloroquinoline 99%
8-Chloroquinoline
Formule: C₉H₆ClN
Kookpunt: 288 °C (1013 hPa)
Smeltpunt: –20 °C
Dichtheid: 1,2834 g/cm³ (14 °C)
Vlampunt: >110 °C
MDL Nummer: MFCD00047618
CAS nummer: 611-33-6
EINECS: 210-265-6
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Residue after evaporation ≤0.001 %
Free amine ≤0.0002 %
Acidity (CH3COOH) ≤0.003 %