Cloxiquina
Leverancier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PROOMM0391.00EA
0
EUR
PROOMM0391.00
Cloxiquina
Cloxiquina
Formule:
C₉H₆ClNO MW: 179,61 g/mol Kookpunt: 349 °C (1013 hPa) Smeltpunt: 125 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006788 CAS nummer: 130-16-5 EINECS: 204-978-1 Merck Index: 12,02483 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...