4-Chloroquinaldine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

BTB04981.1 BTB04981.10 BTB04981.50
MAYBBTB04981.1EA 0 EUR
MAYBBTB04981.1 MAYBBTB04981.10 MAYBBTB04981.50
4-Chloroquinaldine
4-Chloroquinaldine
Formule: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Nummer: MFCD00006757
CAS nummer: 4295-06-1
EINECS: 224-300-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR