6-Nitroquinaldine
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Methyl-6-nitroquinoline
BTB06234DA
BTB06234EA
MAYBBTB06234DAEA
0
EUR
MAYBBTB06234DA
MAYBBTB06234EA
6-Nitroquinaldine
6-Nitroquinaldine
Formule:
C₁₀H₈N₂O₂ |
MDL Nummer:
MFCD00051736 CAS nummer: 613-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...