Bromocyclopentane 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

111600500 111602500 111600010
ACRO111600500EA 21.8 EUR
ACRO111600500 ACRO111602500 ACRO111600010
Bromocyclopentane 98%
Bromocyclopentane
Formule: C₅H₉Br
Kookpunt: 137…139 °C (760 mmHg)
Dichtheid: 1,39 g/cm³
Vlampunt: 35 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001359
CAS nummer: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4881 to 1.4901 (20°C, 589 nm)
GC ≥97.5 %
UV at 260 nm A: ≤0.03
Titration after Ion exchange 98.5 to 101.5 % (on dry substance)
Chloride (Cl) ≤1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR