1-Aminocyclopentanecarboxylic acid ≥97%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 97+%
CAS No.: 52-52-8
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: ca 320° dec.
Beilstein Reference: 636626
Merck Reference: 14,2734
UN No.: 2811
MDL No.: MFCD00001381
Formule:
C₆H₁₁NO₂ Kookpunt: 254 °C (1013 hPa) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001381 CAS nummer: 52-52-8 EINECS: 200-144-6 UN: 2811 ADR: 6.1,III Merck Index: 14,2734 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...