Cyclohexylamine ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A15851.0F
A15851.AE
A15851.AP
A15851.0FEA
67.8
EUR
A15851.0F
A15851.AE
A15851.AP
Cyclohexylamine ≥98%
Cyclohexylamine
Purity: 98+%
CAS No.: 108-91-8
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 27°(81°F)
Refractive Index: 1.4585
Melting Point: -18°
Boiling Point: 133-134°
Beilstein Reference: 471175
Merck Reference: 14,2729
UN No.: 2357
MDL No.: MFCD00001486
Formule:
C₆H₁₃N Kookpunt: 133…134 °C Smeltpunt: –18 °C Dichtheid: 0,868 Vlampunt: 27 °C (81 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001486 CAS nummer: 108-91-8 EINECS: 203-629-0 UN: 2357 ADR: 8,II Merck Index: 14,2729 |