Cyclohexaan, BAKER, Laboratory Reagent, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Hexahydrobenzene, Hexamethylene
8104.1000EA 67.05 EUR
8104.1000 8104.2500
Cyclohexaan, BAKER, Laboratory Reagent, J.T.Baker®
Cyclohexaan
Formule: C₆H₁₂
MW: 84.16 g/mol
Kookpunt: 81
Smeltpunt: 7
Dichtheid: 0.7785 g/cm³ (20 °C)
Vlampunt: –18 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003814
CAS nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use min. 99 %
Residue after Evaporation max. 0.005 %
Water (H2O) max. 0.02 %