5-Bromo-2-furoic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AC10702.1 AC10702.50
MAYBAC10702.1EA 0 EUR
MAYBAC10702.1 MAYBAC10702.50
5-Bromo-2-furoic acid
5-Bromo-2-furoic acid
Formule: C₅H₃BrO₃
MDL Nummer: MFCD00003239
CAS nummer: 585-70-6
EINECS: 209-559-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR