trans-Bis(triphenylphosphine)palladium(II) chloride (≥14,0% Pd)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
trans-Dichlorobis(triphenylphosphine)palladium(II)
10491.03
10491.06
10491.14
10491.03EA
64.6
EUR
10491.03
10491.06
10491.14
trans-Bis(triphenylphosphine)palladium(II) chloride (≥14,0% Pd)
trans-Bis(triphenylphosphine)palladium(II) chloride
CAS No.: 13965-03-2
Molecular Formula: C36H30Cl2P2Pd
Formula Weight: 701.91
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Yellow
Odour: Odourless
Melting Point: ca 310° dec.
MDL No.: MFCD00009593
Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation
Formule:
C₃₆H₃₀Cl₂P₂Pd MW: 701.91 g/mol Smeltpunt: ∼ 310 °C (ontbinding) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009593 CAS nummer: 13965-03-2 EINECS: 237-744-2 |