Lithium trifluoromethanesulphonate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
39322.09
39322.18
39322.09EA
59.9
EUR
39322.09
39322.18
Lithium trifluoromethanesulphonate ≥97%
Lithium trifluoromethanesulphonate
Purity: 97%
CAS No.: 33454-82-9
Molecular Formula: LiCF3SO3
Formula Weight: 156.01
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 423°
MDL No.: MFCD00013227
Formule:
CF₃LiO₃S Smeltpunt: 423 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013227 CAS nummer: 33454-82-9 EINECS: 251-528-5 |