Tin(II) trifluoromethanesulphonate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate
39603.03
39603.06
39603.03EA
49.7
EUR
39603.03
39603.06
Tin(II) trifluoromethanesulphonate ≥97%
Tin(II) trifluoromethanesulphonate
Purity: 97%
CAS No.: 62086-04-8
Molecular Formula: Sn(CF3SO3)2
Formula Weight: 416.82
Physical Form: Powder
Sensitivity: Moisture Sensitive
UN No.: 3288
MDL No.: MFCD00191251
Formule:
C₂F₆O₆S₂Sn MW: 416.85 g/mol Smeltpunt: Dec. Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00191251 CAS nummer: 62086-04-8 UN: 3288 ADR: 6.1 III |