Copper(II) trifluoromethanesulphonate ≥99%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 99% min
CAS No.: 34946-82-2
Molecular Formula: Cu(CF3SO3)2
Formula Weight: 361.68
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to pale blue
Melting Point: dec.
Beilstein Reference: 4028198
UN No.: 3261
MDL No.: MFCD00077492
Formule:
C₂CuF₆O₆S₂ Smeltpunt: Dec. Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00077492 CAS nummer: 34946-82-2 EINECS: 252-300-8 UN: 3261 ADR: 8,III |