2-Chloroethyldimethylammonium chloride ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: N-(2-Chloroethyl)-N,N-Dimethylammonium Chloride, 2-Dimethylaminoethyl chloride hydrochloride
A15134.22 A15134.36
A15134.22EA 27 EUR
A15134.22 A15134.36
2-Chloroethyldimethylammonium chloride ≥98%
2-Chloroethyldimethylammonium chloride
Purity: 98+% CAS No.: 4584-46-7 Molecular Formula: ClCH2CH2N(CH3)2.HCl Formula Weight: 144.05 Melting Point: 205-208° Beilstein Reference: 3591234 UN No.: 2811 MDL No.: MFCD00012516
Formule: C₄H₁₁Cl₂N
MW: 144.04 g/mol
Smeltpunt: 205…208 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012516
CAS nummer: 4584-46-7
EINECS: 224-970-1
UN: 2811
ADR: 6.1 III