Tantalum(V) ethanolate 99.95% Metals basis, Optical Grade Optical Grade
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Tantalum(V) ethoxide
L16033.03 L16033.06
L16033.03EA 27.9 EUR
L16033.03 L16033.06
Tantalum(V) ethanolate 99.95% Metals basis, Optical Grade Optical Grade
Tantalum(V) ethanolate
Purity: 99.95% CAS No.: 6074-84-6 Molecular Formula: C10H25O5Ta Formula Weight: 406.25 Flash Point: 30°(86°F) Refractive Index: 1.4880 Melting Point: 18-19° Boiling Point: 145°/0.1mm Beilstein Reference: 3678999 UN No.: 2924 MDL No.: MFCD00049785
Formule: C₁₀H₂₅O₅Ta
MW: 406.25 g/mol
Kookpunt: 145 °C (0.1 mmHg)
Smeltpunt: 18…19 °C
Dichtheid: 1.566
Vlampunt: 30 °C (86 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049785
CAS nummer: 6074-84-6
EINECS: 228-010-2
UN: 2924
ADR: 3 III