DL-α-Aminoadipic acid

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: DL-2-Aminoadipic acid, (RS)-2-Aminohexanedioic acid, H-DL-Homoglu-OH, 2-Aminohexanedioic acid, H-DL-Aad-OH

4001419.0005 4001419.0025
BACE4001419.0005EA 0 EUR
BACE4001419.0005 BACE4001419.0025
DL-α-Aminoadipic acid
DL-α-Aminoadipic acid
Formule: C₆H₁₁NO₄
MDL Nummer: MFCD00063119
CAS nummer: 542-32-5
EINECS: 208-809-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR