Brij® 35 30% in aqueous solution, Millipore®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Brij® L23
M-Clarity™ quality level = MQ100
1.01894.0100
1.01894.1000
1.01894.0100EA
55.8
EUR
1.01894.0100
1.01894.1000
Brij® 35 30% in aqueous solution, Millipore®
Brij® 35
Formule:
C₅₈H₁₁₈O₂₄ MW: 1199.56 g/mol |
MDL Nummer:
MFCD00080891 CAS nummer: 9002-92-0 EINECS: 500-002-6 |