Cyclohexaan
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
M-Clarity™ quality level = MQ200
1.02830.9190EA
0
EUR
1.02830.9190
Cyclohexaan
Cyclohexaan
Formule:
C₆H₁₂ MW: 84,16 g/mol Kookpunt: 81 °C (1013 hPa) Smeltpunt: 7 °C Dichtheid: 0,7785 g/cm³ (20 °C) Vlampunt: –18 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003814 CAS nummer: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 13,02752 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...