D(-)-Quinic acid

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

8835.1 8835.2
ROTH8835.1EA 0 EUR
ROTH8835.1 ROTH8835.2
D(-)-Quinic acid
D-(-)-Quinic acid
Formule: C₇H₁₂O₆
MDL Nummer: MFCD00003864
CAS nummer: 77-95-2
EINECS: 201-072-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR