4-Propenylveratrole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-(1-Propenyl)pyrocatechol dimethyl ether, 4-(1-Propenyl)-1,2-dimethoxybenzene, 1,2-Dimethoxy-4-Propenylbenzene, 1,2-Dimethoxy-4-(1-propenyl)benzene, Methyl isoeugenol, O-Methyl Isoeugenol
P1103-25ML
P1103-500ML
TCIAP1103-25MLEA
23.1
EUR
TCIAP1103-25ML
TCIAP1103-500ML
4-Propenylveratrole ≥98.0% (by GC)
4-Propenylveratrole
Formule:
C₁₁H₁₄O₂ Kookpunt: 264°C Dichtheid: 1,06 g/cm³ (20 °C) Vlampunt: 100°C |
MDL Nummer:
MFCD00009282 CAS nummer: 93-16-3 EINECS: 202-224-6 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...