Cyclopentaan for HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Pentamethylene
44814.AK
44814.K2
44814.AKEA
59.3
EUR
44814.AK
44814.K2
Cyclopentaan for HPLC
Cyclopentaan
CAS No.: 287-92-3
Molecular Formula: C5H10
Formula Weight: 70.14
Flash Point: -37°(-34°F)
Physical Form: Liquid
Density: 0.748
Refractive Index: 1.4060
Appearance: Colourless
Odour: Petrol-like
Melting Point: -95°
Boiling Point: 47-49°
Beilstein Reference: 1900195
Merck Reference: 14,2741
UN No.: 1146
MDL No.: MFCD00001356
For HPLC and Spectrophotometry
Formule:
C₅H₁₀ Kookpunt: 47…49 °C Smeltpunt: –95 °C Dichtheid: 0,748 Vlampunt: –37 °C (–34 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001356 CAS nummer: 287-92-3 EINECS: 206-016-6 UN: 1146 ADR: 3,II Merck Index: 14,2741 |