2-Heptylcyclopentanone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

SB01363.1 SB01363.10 SB01363.50
MAYBSB01363.1EA 0 EUR
MAYBSB01363.1 MAYBSB01363.10 MAYBSB01363.50
2-Heptylcyclopentanone
2-Heptylcyclopentanone
Formule: C₁₂H₂₂O
MW: 182.31 g/mol
MDL Nummer: MFCD00019315
CAS nummer: 137-03-1
EINECS: 205-273-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR