2-Methylcyclohexanon ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A14741.0F
A14741.AE
A14741.AP
A14741.0FEA
809
EUR
A14741.0F
A14741.AE
A14741.AP
2-Methylcyclohexanon ≥98%
2-Methylcyclohexanon
Purity: 98%
CAS No.: 583-60-8
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 46°(114°F)
Physical Form: Clear liquid
Density: 0.925
Refractive Index: 1.4480
Appearance: Colourless
Melting Point: -14°
Boiling Point: 162-163°
Beilstein Reference: 506751
UN No.: 2297
MDL No.: MFCD00001635
Formule:
C₇H₁₂O Kookpunt: 162…163 °C Smeltpunt: –14 °C Dichtheid: 0,925 Vlampunt: 46 °C (114 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001635 CAS nummer: 583-60-8 EINECS: 209-513-6 UN: 2297 ADR: 3,III |