1-Methylcyclohexanecarboxylic acid
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01941.1 SB01941.10 SB01941.50
MAYBSB01941.1EA 0 EUR
MAYBSB01941.1 MAYBSB01941.10 MAYBSB01941.50
1-Methylcyclohexanecarboxylic acid
1-Methylcyclohexanecarboxylic acid
Formule: C₈H₁₄O₂
Kookpunt: 234 °C (1013 hPa)
Smeltpunt: 39 °C
Dichtheid: 1,0115 g/cm³ (20 °C)
MDL Nummer: MFCD00001463
CAS nummer: 1123-25-7
EINECS: 214-371-3