1-Methylcyclohexanol 96%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
126600050 126600250 126601000
ACRO126600050EA 21.9 EUR
ACRO126600050 ACRO126600250 ACRO126601000
1-Methylcyclohexanol 96%
1-Methylcyclohexanol
Formule: C₇H₁₄O
Kookpunt: 168 °C (752 mmHg)
Smeltpunt: 26 °C
Dichtheid: 0,919 g/cm³
Vlampunt: 67 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003857
CAS nummer: 590-67-0
EINECS: 215-963-4
UN: 1325
ADR: 4.1,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20°C, 589 nm)
GC ≥95 %