1-Methylcyclopentanol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B24744.06 B24744.22
B24744.06EA 54.2 EUR
B24744.06 B24744.22
1-Methylcyclopentanol ≥98%
1-Methylcyclopentanol

Purity: 98% CAS No.: 1462-03-9 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 40°(105°F) Melting Point: 36-37° Boiling Point: 135-136° UN No.: 1325 MDL No.: MFCD00001364

Formule: C₆H₁₂O
Kookpunt: 135…136 °C
Smeltpunt: 36…37 °C
Dichtheid: 0,904
Vlampunt: 40 °C (105 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001364
CAS nummer: 1462-03-9
EINECS: 215-963-4
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR