Methylcyclopentane for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.20789.0005 8.20789.0250
8.20789.0005EA 31.4 EUR
8.20789.0005 8.20789.0250
Methylcyclopentane for synthesis, Sigma-Aldrich®
Methylcyclopentane
Formule: C₆H₁₂
Kookpunt: 71,8 °C (1013 hPa)
Smeltpunt: –142 °C
Dichtheid: 0,74 g/cm³ (20 °C)
Vlampunt: –17 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001382
CAS nummer: 96-37-7
EINECS: 202-503-2
UN: 2298
ADR: 3,II
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -142 °C
Molar Mass 84.16 g/mol
Boiling Point 72 °C (1013 mbar)
Vapor Pressure 150 hPa (20 °C)
Flash Point -17 °C
Refractive Index 1.4087 (20 °C, 589 nm)
Density 0.74 g/cm3 (20 °C)
Ignition Point 315 °C