Pentylcyclohexane ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
B21326.04
B21326.09
B21326.04EA
48.7
EUR
B21326.04
B21326.09
Pentylcyclohexane ≥98%
Pentylcyclohexane
Purity: 98%
CAS No.: 4292-92-6
Molecular Formula: C11H22
Formula Weight: 154.30
Flash Point: 63°(145°F)
Refractive Index: 1.4445
Odour: Mild
Melting Point: -58°
Boiling Point: 202°
UN No.: 3082
MDL No.: MFCD00019402
Formule:
C₁₁H₂₂ Kookpunt: 202 °C Smeltpunt: –58 °C Dichtheid: 0,800 Vlampunt: 63 °C (145 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00019402 CAS nummer: 4292-92-6 EINECS: 224-296-8 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...