Cycloheptene 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
304260050 304260250
ACRO304260050EA 97.8 EUR
ACRO304260050 ACRO304260250
Cycloheptene 95%
Cycloheptene
Formule: C₇H₁₂
Kookpunt: 114,6 °C (1013 hPa)
Smeltpunt: –56 °C
Dichtheid: 0,82 g/cm³
Vlampunt: –006 °C
MDL Nummer: MFCD00004156
CAS nummer: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4610 (20°C, 589 nm)
GC ≥94 %
Stabilizer 0.1 % BHT