2-Bromofluorene 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L07589.06
L07589.14
L07589.06EA
37.8
EUR
L07589.06
L07589.14
2-Bromofluorene 95%
2-Bromofluorene
Purity: 95%
CAS No.: 1133-80-8
Molecular Formula: C13H9Br
Formula Weight: 245.12
Melting Point: 110-114°
Boiling Point: 185°/135mm
MDL No.: MFCD00001115
Formule:
C₁₃H₉Br MW: 245.12 g/mol Kookpunt: 185 °C (135 mmHg) Smeltpunt: 110…114 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001115 CAS nummer: 1133-80-8 EINECS: 214-480-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...