Sodium borohydride 98+%, powder
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Sodium tetrahydridoborate, Sodium tetrahydroborate
189300050 189300100 189301000 189305000 189300025
ACRO189300050EA 29.3 EUR
ACRO189300050 ACRO189300100 ACRO189301000 ACRO189305000 ACRO189300025
Sodium borohydride 98+%, powder
Sodium borohydride
Formule: NaBH₄
MW: 37.83 g/mol
Smeltpunt: 360 °C (ontbinding)
Vlampunt: 69
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003518
CAS nummer: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664
Order Now
Specification Test Results
Appearance White Powder
Titration Iodometric ≥98.0 %
Water ≤5 %
HPLC ≥98.5 %