Potassium bromide ≥99.92% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
10982.14
10982.22
10982.14EA
75.7
EUR
10982.14
10982.22
Potassium bromide ≥99.92% (metals basis)
Potassium bromide
Purity: 99.92%
CAS No.: 7758-02-3
Molecular Formula: KBr
Formula Weight: 119.01
Physical Form: Crystalline
Density: 2.75
Refractive Index: 1.559
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 730°
Boiling Point: 1435°
Merck Reference: 14,7618
MDL No.: MFCD00011358
Formule:
KBr Kookpunt: 1435 °C Smeltpunt: 730 °C Dichtheid: 2,75 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011358 CAS nummer: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |