Sodium bromide, (max. 1% H₂O) 99+% (dry weight)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A10552.0B A10552.0I A10552.30
A10552.0BEA 59 EUR
A10552.0B A10552.0I A10552.30
Sodium bromide, (max. 1% H₂O) 99+% (dry weight)
Sodium bromide
Purity: 99+% CAS No.: 7647-15-6 Molecular Formula: NaBr Formula Weight: 102.89 Refractive Index: 1.6412 Melting Point: 755° Boiling Point: 1390° Merck Reference: 14,8594 MDL No.: MFCD00003475
Formule: NaBr
Kookpunt: 1390 °C
Smeltpunt: 755 °C
Dichtheid: 3,203
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003475
CAS nummer: 7647-15-6
EINECS: 231-599-9
Merck Index: 14,8594