Sodium bromate 99+%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
208871000 208875000 208870025
ACRO208871000EA 28.8 EUR
ACRO208871000 ACRO208875000 ACRO208870025
Sodium bromate 99+%, extra pure
Sodium bromate
Formule: BrNaO₃
MW: 150.89 g/mol
Kookpunt: 1390
Smeltpunt: 381 °C (ontbinding)
Dichtheid: 3.33 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003476
CAS nummer: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1 II
Merck Index: 12,08736
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Iodometric ≥99.0 %
Bromide (Br) ≤500 ppm
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm