Potassium carbonate, anhydrous 99+% ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Potash
424085000 424080025 424080100 424081000
ACRO424085000EA 55.2 EUR
ACRO424085000 ACRO424080025 ACRO424080100 ACRO424081000
Potassium carbonate, anhydrous 99+% ACS
Potassium carbonate
Formule: K₂CO₃
MW: 138.21 g/mol
Smeltpunt: 891 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011382
CAS nummer: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Granular powder
Titration with HCl ≥99.0 %
Chloride (Cl) ≤0.003 %
Phosphate (PO4) ≤0.001 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.02 %
Heavy metals (ICP-OES) ≤5 ppm
Silicon dioxide (SiO2) ≤0.005 %
Sulfur compounds (as SO4) ≤0.004 %
Insoluble matter ≤0.01 %