Potassium carbonate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P743.1 P743.2
ROTHP743.1EA 42.5 EUR
ROTHP743.1 ROTHP743.2
Potassium carbonate
Potassium carbonate
Formule: K₂CO₃
Smeltpunt: 891 °C
Dichtheid: 2,42 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011382
CAS nummer: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR