Potassium carbonate
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Potash
X894.1
X894.2
X894.3
ROTHX894.1EA
23.9
EUR
ROTHX894.1
ROTHX894.2
ROTHX894.3
Potassium carbonate
Potassium carbonate
Formule:
K₂CO₃ Smeltpunt: 891 °C Dichtheid: 2,42 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011382 CAS nummer: 584-08-7 EINECS: 209-529-3 Merck Index: 13,07702 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...