Lithium carbonate ≥99.998% (metals basis excluding Ca), Puratronic® (max. 0.002% Ca)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
10734.18
10734.30
10734.A1
10734.18EA
116
EUR
10734.18
10734.30
10734.A1
Lithium carbonate ≥99.998% (metals basis excluding Ca), Puratronic® (max. 0.002% Ca)
Lithium carbonate
Purity: 99.998%
CAS No.: 554-13-2
Molecular Formula: Li2CO3
Formula Weight: 73.90
Physical Form: Powder
Density: 2.11
Appearance: White
Odour: Odourless
pH: 10-11.5
Melting Point: 723°
Boiling Point: 1310° dec.
Merck Reference: 14,5527
MDL No.: MFCD00011084
Formule:
Li₂CO₃ MW: 73.89 g/mol Kookpunt: 1310 °C (ontbinding) Smeltpunt: 723 °C Dichtheid: 2.11 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011084 CAS nummer: 554-13-2 EINECS: 209-062-5 Merck Index: 14,5527 |