Lithium carbonate, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra pure suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
1.05671.1000 1.05671.9050
1.05671.1000EA 1230 EUR
1.05671.1000 1.05671.9050
Lithium carbonate, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra pure suitable for use as excipient, SAFC®
Lithium carbonate
Formule: Li₂CO₃
MW: 73.89 g/mol
Smeltpunt: 720
Dichtheid: 2.11 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011084
CAS nummer: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549