Evaporating dishes, conical, with spout, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A169-03 A169-04 A169-02 A169-01
539-1415EA 47 EUR
539-1415 BOHLA169-04 BOHLA169-02 BOHLA169-01
Evaporating dishes, conical, with spout, BOLA
Reservoirs/Schalen/Potten Uitdampschalen
Made of PTFE, white.
  • Universal chemical resistance
  • Temperature resistance: from −200 to +250 °C