Magnetic Stirring Bars, glass, cylindrical, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C351-19 C351-15 C351-12 C351-09 C351-06 C351-03
BOHLC351-19EA 23.9 EUR
BOHLC351-19 BOHLC351-15 BOHLC351-12 BOHLC351-09 BOHLC351-06 BOHLC351-03
Magnetic Stirring Bars, glass, cylindrical, BOLA
Roerders Magnetische roerstaven
Cylindrical magnetic stirring bars with Alnico V magnetic core encapsulated in borosilicate glass.
  • Non porous, non contaminating, universal chemical resistance
  • For high-purity work or trace analysis
Cylindrical shape with extremely smooth surface prevents penetration of substances.
Order Now
Specificaties
Environmentally Preferable