Magnetic stirring bars, PTFE, triangular, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C357-03 C357-09 C357-15 C357-21 C357-30 C357-06 C357-12 C357-18 C357-27 C357-24
BOHLC357-03EA 4.6 EUR
BOHLC357-03 BOHLC357-09 BOHLC357-15 BOHLC357-21 BOHLC357-30 BOHLC357-06 BOHLC357-12 BOHLC357-18 BOHLC357-27 BOHLC357-24
Magnetic stirring bars, PTFE, triangular, BOLA
Roerders Magnetische roerstaven
Triangular shaped magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE.
  • Chemical resistant
Temperature resistance: −200 to +250 °C.
Most commonly used for big vessels in which strong turbulence can be acheived with low speeds. These can also been useful for mixing of reagents which resist dissolving and for avoiding any residues at the bottom of the vessels.