Magnetic stirring bars, PTFE, egg-shaped, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C358-02 C358-06 C358-10 C358-12 C358-08 C358-04 C358-16 C358-14
BOHLC358-02EA 5.1 EUR
BOHLC358-02 BOHLC358-06 BOHLC358-10 BOHLC358-12 BOHLC358-08 BOHLC358-04 BOHLC358-16 BOHLC358-14
Magnetic stirring bars, PTFE, egg-shaped, BOLA
Roerders Magnetische roerstaven
Egg shaped magnetic stirrer bars with magnetic core of Alnico 5 magnet encapsulated in PTFE. The shape mimics that of the flask and ensures complete mixing.
  • Chemical resistant
Temperature resistance: −200 to +250 °C.