Magnetic stirring bars, PTFE, double-ended, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C359-03 C359-06
BOHLC359-03EA 23.9 EUR
BOHLC359-03 BOHLC359-06
Magnetic stirring bars, PTFE, double-ended, BOLA
Roerders Magnetische roerstaven
Double end shaped PTFE magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE. Stable discs present at both sides provide an excellent stirring capability.
  • Chemical resistant
Temperature resistance: −200 to +250 °C.