Bearing neck for tandem magnetic stirring bar, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C364-16 C364-08
BOHLC364-16EA 27 EUR
BOHLC364-16 BOHLC364-08
Bearing neck for tandem magnetic stirring bar, BOLA
Roerders Roergereedschap
Bearing necks for receiving tandem magnetic stirring bar. The centre portion will fix to the vessel by means of glue such as silicone hardened in water.
  • Chemical resistance
  • Made of very hard PTFE/PEEK compound
Temperature resistance: −100 to +240 °C.