Magnetic stirring bars, centred, PTFE-encapsulated, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C367-50 C367-40 C367-30 C367-20
BOHLC367-50EA 10.2 EUR
BOHLC367-50 BOHLC367-40 BOHLC367-30 BOHLC367-20
Magnetic stirring bars, centred, PTFE-encapsulated, BOLA
Roerders Magnetische roerstaven
PTFE-encapsulated magnetic core (Alnico 5), conically tapered ends.
  • Chemical resistant
  • Extremely steady mixing due to central magnet
Temperature resistance: −200 to +250 °C.
Order Now
Specificaties
Environmentally Preferable