Potassium hydrogen carbonate, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, E501, FCC suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Potassium bicarbonate
1.04852.9029 1.04852.9500 1.04852.1000
1.04852.9029EA 0 EUR
1.04852.9029 1.04852.9500 1.04852.1000
Potassium hydrogen carbonate, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, E501, FCC suitable for use as excipient, SAFC®
Potassium hydrogen carbonate
Formule: KHCO₃
MW: 100.12 g/mol
Smeltpunt: 292
Dichtheid: 2.17 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011402
CAS nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691