Sodium hydrogen carbonate, EMPROVE® EXPERT JP, USP, BP, Ph. Eur. for biopharmaceutical production, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium bicarbonate
1.37013.2500 1.37013.9012 1.37013.9029
1.37013.2500EA 94.8 EUR
1.37013.2500 1.37013.9012 1.37013.9029
Sodium hydrogen carbonate, EMPROVE® EXPERT JP, USP, BP, Ph. Eur. for biopharmaceutical production, SAFC®
Sodium hydrogen carbonate
Formule: NaHCO₃
Smeltpunt: 270 °C
Dichtheid: 2,159 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003528
CAS nummer: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM