Sodium hydrogen carbonate

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Sodium bicarbonate

39780.01 39780.02
SERA39780.01EA 0 EUR
SERA39780.01 SERA39780.02
Sodium hydrogen carbonate
Sodium hydrogen carbonate
Formule: NaHCO₃
Smeltpunt: 270 °C
Dichtheid: 2,159 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003528
CAS nummer: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR