Potassium chloride 99,997 Metals basis, Puratronic®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Synoniemen:
Potassiumchloride
10839.18
10839.30
10839.18EA
159
EUR
10839.18
10839.30
Potassium chloride 99,997 Metals basis, Puratronic®
Potassium chloride
Purity: 99.997%
CAS No.: 7447-40-7
Molecular Formula: KCl
Formula Weight: 74.55
Physical Form: Powder
Density: 1.984
Refractive Index: 1.490
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 773°
Boiling Point: 1500° subl.
Merck Reference: 14,7621
MDL No.: MFCD00011360
Formule:
KCl MW: 74.55 g/mol Kookpunt: 1500 °C (sublimatie) Smeltpunt: 773 Dichtheid: 1.984 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011360 CAS nummer: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,7621 |